Learn More
4-Nitrobenzenemethanol, TRC
Product Code. 30474169
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474169 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474169 Supplier Toronto Research Chemicals Supplier No. TRCN493520100G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Nitrobenzenemethanol
CAS 619-73-8
Molecular Formula C7H7NO3
Quantity 100 g
Synonym (4-Nitrophenyl)methanol, 1-(Hydroxymethyl)-4-nitrobenzene, 4-(Hydroxymethyl)nitrobenzene, 4-Nitrobenzenemethanol, 4-Nitrobenzyl alcohol, NSC 5389, p-(Hydroxymethyl)nitrobenzene, p-Nitrobenzyl alcohol, p-Nitrophenylmethanol, p-Nitrobenzyl Alcohol
InChI Formula InChI=1S/C7H7NO3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4,9H,5H2
SMILES OCc1ccc(cc1)[N+](=O)[O-]
IUPAC Name (4-nitrophenyl)methanol
Molecular Weight (g/mol) 153.14
Formula Weight 153.0426