Learn More
4-Nitrophenyl dodecanoate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1956-11-2 |
| Molecular Formula | C18H27NO4 |
| MDL Number | MFCD00047730 |
| Quantity | 5 g |
| SMILES | CCCCCCCCCCCC(OC1=CC=C([N+]([O-])=O)C=C1)=O |
| Molecular Weight (g/mol) | 321.41 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Nitrophenyl dodecanoate |