Learn More
4-Phenoxyphenol, TRC
Product Code. 30417037
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417037 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417037 Supplier Toronto Research Chemicals Supplier No. TRCP301380100G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Phenoxyphenol
CAS 831-82-3
Molecular Formula C12 H10 O2
Quantity 100 g
Synonym Phenol','p-phenoxy- (6CI,7CI,8CI), 4-Phenoxyphenol, 4-Hydroxydiphenyl ether, Hydroquinone monophenyl ether, NSC 25027, p-Hydroxydiphenyl ether, p-Phenoxyphenol, 4-Hydroxydiphenyl Ether, Hydroquinone Monophenyl Ether, NSC 25027, p-Hydroxydiphenyl Ether, p-Phenoxyphenol, Phenol','p-phenoxy- (6CI,7CI,8CI)
InChI Formula InChI=1S/C12H10O2/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9,13H
SMILES Oc1ccc(Oc2ccccc2)cc1
IUPAC Name 4-phenoxyphenol
Molecular Weight (g/mol) 186.21
Formula Weight 186.0681