Learn More
4-Phenyloxolan-2-one, TRC
Product Code. 30414074
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414074 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414074 Supplier Toronto Research Chemicals Supplier No. TRCP32151810MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Phenyloxolan-2-one
CAS 1008-73-7
Molecular Formula C10H10O2
Quantity 10 mg
InChI Formula InChI=1S/C10H10O2/c11-10-6-9(7-12-10)8-4-2-1-3-5-8/h1-5,9H,6-7H2
SMILES C1C(COC1=O)C2=CC=CC=C2
IUPAC Name 4-phenyloxolan-2-one
Molecular Weight (g/mol) 162.18
Formula Weight 162.0681