Learn More
Labconco 4-Port Manifold for CT-Series Freeze Dryer
Product Code. 31703094 Shop All Labconco Corporation Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
31703094 -50°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31703094 Supplier Labconco Supplier No. 7232800
  • ONLINE EXCLUSIVE PRICE
    1180.00€
    931.23€ / 1 pièces
    Économisez 248.77€
    21% de réduction
 Restricted Item
Expédition estimée: 26-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Manifold provides easy connection for adapters and flasks.
  • Manifold with four neoprene valves with plastic knobs that accommodate both 1/2 in. and 3/4 in. adapters for connection of flask. For use with Labconco CT Series Freeze Dryers.
Specifications
Type Drying Manifold
No. of Shelves 0
Temperature Range (Metric) -50°C
Material Stainless Steel
Plug Type All except North America
Voltage 0/1 V