Learn More
4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-[2,2'-bipyrimidin]-4-yl)benzenesulfonamide hydrate, 98%, BLD Pharm™
Product Code. 30777118
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30777118 1 g 1g
30771919 100 mg 100mg
30773990 5 g 5g
30772613 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30777118 Supplier BLD Pharm Supplier No. BD1186441g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 157212-55-0
Molecular Formula C27H31N5O7S
MDL Number MFCD09751188
Quantity 1 g
SMILES O=S(C1=CC=C(C(C)(C)C)C=C1)(NC2=NC(C3=NC=CC=N3)=NC(OCCO)=C2OC4=CC=CC=C4OC)=O.[H]O[H]
Molecular Weight (g/mol) 569.63
Percent Purity 98%
Chemical Name or Material 4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-[2,2'-bipyrimidin]-4-yl)benzenesulfonamide hydrate