Learn More
[5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol, TRC
Product Code. 30472731
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472731 50 mg 50mg
30466118 500 mg 500mg
30474586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30472731 Supplier Toronto Research Chemicals Supplier No. TRCM69572050MG
 Restricted Item
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 337508-70-0
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.3
IUPAC Name [5-(2-methyl-1,3-thiazol-4-yl)thiophen-2-yl]methanol
SMILES Cc1nc(cs1)c2ccc(CO)s2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C9H9NOS2/c1-6-10-8(5-12-6)9-3-2-7(4-11)13-9/h2-3,5,11H,4H2,1H3
Formula Weight 211.0126
Chemical Name or Material [5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol