Learn More
5-(2-Pyridinylsulfanyl)-2-furaldehyde, 95+%, Thermo Scientific™
Product Code. 10432043
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10432043 1 g 1g
10071524 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10432043 Supplier Thermo Scientific Maybridge Supplier No. MO07099DA
 Restricted Item
Expédition estimée: 26-11-2026
Add to basket
This item is not returnable. View return policy
Chemical Identifiers
CAS 709635-68-7
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.231
InChI Key NWJZBQDKKLHSBP-UHFFFAOYSA-N
Synonym 5-2-pyridinylsulfanyl-2-furaldehyde, 5-pyridin-2-ylthio furan-2-carbaldehyde, 5-pyridin-2-ylsulfanyl furan-2-carbaldehyde, 5-pyridin-2-ylsulfanyl-furan-2-carbaldehyde, 5-pyridin-2-ylthio-2-furaldehyde, 5-2-pyridylthio furan-2-carbaldehyde, 5-2-pyridylsulfanyl furan-2-carbaldehyde, 2-furancarboxaldehyde,5-2-pyridinylthio, 2-furancarboxaldehyde, 5-2-pyridinylthio
PubChem CID 2794784
IUPAC Name 5-pyridin-2-ylsulfanylfuran-2-carbaldehyde
SMILES C1=CC=NC(=C1)SC2=CC=C(O2)C=O
Specifications
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 205.24
Percent Purity 95%
Chemical Name or Material 5-(2-Pyridinylsulfanyl)-2-furaldehyde