Learn More
5,6-Dimethoxy-2-[(4-piperidyl)methyl]indane Hydrochloride(Donepezil Impurity), TRC
Product Code. 30368997
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368997 Supplier Toronto Research Chemicals Supplier No. TRCD46072150MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5,6-Dimethoxy-2-[(4-piperidyl)methyl]indane Hydrochloride(Donepezil Impurity)
CAS 1034439-43-4
Molecular Formula C17H26ClNO2
Quantity 50 mg
Synonym 4-[(2,3-Dihydro-5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine Hydrochloride, Donepezil Desbenzyl Deoxy Impurity F
InChI Formula InChI=1S/C17H25NO2.ClH/c1-19-16-10-14-8-13(7-12-3-5-18-6-4-12)9-15(14)11-17(16)20-2;/h10-13,18H,3-9H2,1-2H3;1H
SMILES Cl.COc1cc2CC(CC3CCNCC3)Cc2cc1OC
Molecular Weight (g/mol) 311.85
Formula Weight 311.1652