Learn More
5-Amino-1-naphthol, TRC
Product Code. 30319670
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319670 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319670 Supplier Toronto Research Chemicals Supplier No. TRCA6186701G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Amino-1-naphthol
CAS 83-55-6
Molecular Formula C10H9NO
Quantity 1 g
Synonym 1-Amino-5-hydroxynaphthalene, 1-Amino-5-naphthol, 5-Amino-1-naphthalenol, 5-Amino-1-naphthol, 5-Amino-α-naphthol, 5-Hydroxy-1-naphthalenamine, 5-Hydroxy-1-naphthylamine, C.I. 76650, NSC 1499
InChI Formula InChI=1S/C10H9NO/c11-9-5-1-4-8-7(9)3-2-6-10(8)12/h1-6,12H,11H2
SMILES OC1=C2C=CC=C(N)C2=CC=C1
IUPAC Name 5-aminonaphthalen-1-ol
Molecular Weight (g/mol) 159.18
Formula Weight 159.0684