Learn More
5-Amino-1-naphthol, TRC
Product Code. 30317103
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317103 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317103 Supplier Toronto Research Chemicals Supplier No. TRCA618670250MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Amino-1-naphthol
CAS 83-55-6
Molecular Formula C10H9NO
Quantity 250 mg
Synonym 1-Amino-5-hydroxynaphthalene, 1-Amino-5-naphthol, 5-Amino-1-naphthalenol, 5-Amino-1-naphthol, 5-Amino-α-naphthol, 5-Hydroxy-1-naphthalenamine, 5-Hydroxy-1-naphthylamine, C.I. 76650, NSC 1499
InChI Formula InChI=1S/C10H9NO/c11-9-5-1-4-8-7(9)3-2-6-10(8)12/h1-6,12H,11H2
SMILES OC1=C2C=CC=C(N)C2=CC=C1
IUPAC Name 5-aminonaphthalen-1-ol
Molecular Weight (g/mol) 159.18
Formula Weight 159.0684