missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4H-1,2,4-triazole-3-thiol, tech., Thermo Scientific™

Product Code. 10196962
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
Product Code. Quantity unitSize
10196962 1 g 1g
10669724 250 mg 250mg
10596995 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 10196962

Brand: Thermo Scientific Maybridge SB00513DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16691-43-3
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.14
MDL Number MFCD00005231
InChI Key WZUUZPAYWFIBDF-UHFFFAOYSA-N
Synonym 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
PubChem CID 2723869
IUPAC Name 5-amino-1,2-dihydro-1,2,4-triazole-3-thione
SMILES NC1=NC(=S)NN1

Specifications

Quantity 1 g
Formula Weight 116.15
Chemical Name or Material 5-Amino-4H-1, 2, 4-triazole-3-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.