Learn More
5-Benzyloxyindole-3-carboxaldehyde, 98%, Thermo Scientific™
Product Code. 10189739 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10189739 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10189739 Supplier Thermo Scientific Acros Supplier No. 433430010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Specifications
CAS 6953-22-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 241°C to 242°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C16H13NO2
MDL Number MFCD00014562
Quantity 1 g
Synonym 5-benzyloxyindole-3-carboxaldehyde, 5-benzyloxyindole-3-carbaldehyde, 5-benzyloxy-1h-indole-3-carbaldehyde, 5-phenylmethoxy-1h-indole-3-carbaldehyde, 5-benzyloxy-3-formylindole, 5-benzyloxy-indole-3-carboxaldehyde, 5-benzyloxy-3-formyl-1h-indole, 5-benzyloxy-3-indolecarboxaldehyde, 5-benzyloxy indole-3-carboxyaldehyde
InChI Key DJGNUBADRQIDNQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3C=O
IUPAC Name 5-phenylmethoxy-1H-indole-3-carbaldehyde
Molecular Weight (g/mol) 251.28
PubChem CID 81398
Formula Weight 251.28
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyloxyindole-3-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 5-Benzyloxyindole-3-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.