Learn More
5-Bromo-2-fluorobenzonitrile, TRC
Product Code. 30341863
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341863 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341863 Supplier Toronto Research Chemicals Supplier No. TRCB6920351G
 Restricted Item
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Bromo-2-fluorobenzonitrile
CAS 179897-89-3
Molecular Formula C7H3BrFN
Quantity 1 g
Synonym Benzonitrile, 5-bromo-2-fluoro- (9CI, ACI), 5-Bromo-2-fluorobenzonitrile (ACI), 2-Fluoro-5-bromobenzonitrile, 3-Bromo-6-fluorobenzonitrile, 3-Cyano-4-fluorobromobenzene
InChI Formula InChI=1S/C7H3BrFN/c8-6-1-2-7(9)5(3-6)4-10/h1-3H
SMILES Fc1ccc(Br)cc1C#N
IUPAC Name 5-bromo-2-fluorobenzonitrile
Molecular Weight (g/mol) 200.01
Formula Weight 198.9433