Learn More
5-Bromosalicylaldehyde, TRC
Product Code. 30330807
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330807 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330807 Supplier Toronto Research Chemicals Supplier No. TRCB68697525G
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Bromosalicylaldehyde
CAS 1761-61-1
Molecular Formula C7H5BrO2
Quantity 25 g
Synonym 5-Bromo-2-hydroxy-benzaldehyde, 5-Bromo-2-hydroxybenzaldehyde, NSC 7310, NSC 9258
InChI Formula InChI=1S/C7H5BrO2/c8-6-1-2-7(10)5(3-6)4-9/h1-4,10H
SMILES Oc1ccc(Br)cc1C=O
IUPAC Name 5-bromo-2-hydroxybenzaldehyde
Molecular Weight (g/mol) 201.02
Formula Weight 199.9473