Learn More
5-Chloro-2-furaldehyde, TRC
Product Code. 30354329
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30354329 100 mg
30353266 500 mg
30360371 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354329 Supplier Toronto Research Chemicals Supplier No. TRCC381980100MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21508-19-0
Molecular Formula C5H3O2Cl
Molecular Weight (g/mol) 130.52
IUPAC Name 5-chlorofuran-2-carbaldehyde
SMILES Clc1oc(C=O)cc1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C5H3ClO2/c6-5-2-1-4(3-7)8-5/h1-3H
Formula Weight 129.9822
Chemical Name or Material 5-Chloro-2-furaldehyde