Learn More
5-(Dimethylamino)naphthalene-1-sulfonic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4272-77-9 |
| Molecular Formula | C12H13NO3S |
| MDL Number | MFCD00149264 |
| Quantity | 1 g |
| SMILES | O=S(C1=C2C=CC=C(N(C)C)C2=CC=C1)(O)=O |
| Molecular Weight (g/mol) | 251.30 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-(Dimethylamino)naphthalene-1-sulfonic acid |