Learn More
5-Hydroxybenzo[de]isochromene-1,3-dione, 95%, BLD Pharm™
Product Code. 30947131
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30947131 5 g 5g
30945643 250 mg 250mg
30946097 1 g 1g
30937983 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30947131 Supplier BLD Pharm Supplier No. BD143415g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 23204-36-6
Molecular Formula C12H6O4
MDL Number MFCD00059759
Quantity 5 g
SMILES O=C(C1=CC=CC2=CC(O)=CC3=C12)OC3=O
Molecular Weight (g/mol) 214.17
Percent Purity 95%
Chemical Name or Material 5-Hydroxybenzo[de]isochromene-1,3-dione