missing translation for 'onlineSavingsMsg'
Learn More

5-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™

Product Code. 10136733
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10136733 1 g Amber Glass Bottle 1g
10522334 250 mg Amber Glass Bottle 250mg
2 options
This item is not returnable. View return policy

Product Code. 10136733

Brand: Thermo Scientific Maybridge CC41406DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-16-8
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD09702358
InChI Key RDQRMCJCROUHOF-UHFFFAOYSA-N
PubChem CID 24229489
SMILES CN1C=CC2=CC(=CC=C12)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-isocyanato-1-methyl-1h-indole, 1h-indole, 5-isocyanato-1-methyl, 1-methylindol-5-isocyanate, 1h-indole,5-isocyanato-1-methyl, 1-methyl-1h-indole-5-yl isocyanate
IUPAC Name 5-isocyanato-1-methyl-1H-indole
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 5-Isocyanato-1-methyl-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.