Learn More
5-Methyl-3-heptanone, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Methyl-3-Heptanone |
| CAS | 541-85-5 |
| Molecular Formula | C8 H16 O |
| Quantity | 50 mL |
| Synonym | 3-Heptanone','5-methyl-, 5-Methyl-3-heptanone, (±)-5-Methyl-3-heptanone, 2-Methylbutyl ethyl ketone, 3-Methyl-5-heptanone, Ethyl 2-methylbutyl ketone, NSC 100734 |
| InChI Formula | InChI=1S/C8H16O/c1-4-7(3)6-8(9)5-2/h7H,4-6H2,1-3H3 |
| SMILES | CCC(C)CC(=O)CC |
| IUPAC Name | 5-methylheptan-3-one |
| Molecular Weight (g/mol) | 128.21 |
| Formula Weight | 128.1201 |