Learn More
5-Methylcytidine, 98%, Thermo Scientific™
Product Code. 17730629 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17730629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17730629 Supplier Thermo Scientific Acros Supplier No. 229000010
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy
Specifications
Molecular Weight (g/mol) 257.25
Shipping Condition No special requirements
Purity ≥97.5%
Quantity 1 g
Solubility Information Soluble aqueous solution
Color White to Light Yellow