Learn More
5-Methylnicotinamide, TRC
Product Code. 30341258
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341258 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341258 Supplier Toronto Research Chemicals Supplier No. TRCB49530850MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methylnicotinamide
CAS 70-57-5
Molecular Formula C7H8N2O
Quantity 50 mg
Synonym NSC 87668, 5-Methyl-3-pyridinecarboxamide
InChI Formula InChI=1S/C7H8N2O/c1-5-2-6(7(8)10)4-9-3-5/h2-4H,1H3,(H2,8,10)
SMILES Cc1cncc(c1)C(=O)N
IUPAC Name 5-methylpyridine-3-carboxamide
Molecular Weight (g/mol) 136.15
Formula Weight 136.0637