Learn More
5-Methylpyrimidine, 97+%, Thermo Scientific™
Product Code. 11355435
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355435 250 mg 250mg
11345435 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355435 Supplier Thermo Scientific Alfa Aesar Supplier No. L19638.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2036-41-1
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00047386
InChI Key TWGNOYAGHYUFFR-UHFFFAOYSA-N
Synonym pyrimidine, 5-methyl, 5-methyl-pyrimidine, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, 5-methyl-pyrimidin, pubchem7207, rarechem am la 0031, 5-methyl-1,3-diazine, acmc-1cnd2, 5-methylpyrimidine, 5-methylpyrimidine, 97+%
PubChem CID 74859
IUPAC Name 5-methylpyrimidine
SMILES CC1=CN=CN=C1
Specifications
Melting Point 29°C to 31°C
Boiling Point 152°C to 154°C
Flash Point 53°C (127°F)
Quantity 250 mg
UN Number UN1325
Beilstein 105684
Formula Weight 94.12
Percent Purity ≥97%
Chemical Name or Material 5-Methylpyrimidine