Learn More
6-(3-Chloropropylamino)-1,3-dimethyl-2,4(1H,3H)-pyrimidinedione, TRC
Product Code. 30134561
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134561 10 g 10g
30012390 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134561 Supplier Toronto Research Chemicals Supplier No. TRCC37980010G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34654-81-4
Molecular Formula C9H14ClN3O2
Molecular Weight (g/mol) 231.68
Synonym 6-[(3-Chloropropyl)amino]-1,3-dimethyl-2,4(1H,3H)-pyrimidinedione, 6-[(3-Chloropropyl)amino]-1,3-dimethyluracil;
IUPAC Name 6-(3-chloropropylamino)-1,3-dimethylpyrimidine-2,4-dione
SMILES CN1C(=O)C=C(NCCCCl)N(C)C1=O
Specifications
Quantity 10 g
InChI Formula InChI=1S/C9H14ClN3O2/c1-12-7(11-5-3-4-10)6-8(14)13(2)9(12)15/h6,11H,3-5H2,1-2H3
Formula Weight 231.0775
Chemical Name or Material 6-(3-Chloropropylamino)-1,3-dimethyl-2,4(1H,3H)-pyrimidinedione