Learn More
6,8-Dibromoimidazo[1,2-a]pyrazine, 97%, BLD Pharm™
Product Code. 31431982
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31431982 5 g
31420984 100 g
31426020 500 g
31424190 25 g
31419547 1 g
31420489 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31431982 Supplier BLD Pharm Supplier No. BD1079185g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 63744-22-9
Molecular Formula C6H3Br2N3
MDL Number MFCD08460056
Quantity 5 g
SMILES BrC1=NC(Br)=CN2C1=NC=C2
Molecular Weight (g/mol) 276.92
Percent Purity 97%
Chemical Name or Material 6,8-Dibromoimidazo[1,2-a]pyrazine