Learn More
6-Amino-5-bromoquinoxaline, TRC
Product Code. 17962095
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962095 1 g 1g
30134380 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17962095 Supplier Toronto Research Chemicals Supplier No. TRCA6018851G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 50358-63-9
Molecular Formula C8 H6 Br N3
Molecular Weight (g/mol) 224.06
Synonym 5-Bromo-6-quinoxalinamine, 5-Bromo-6-aminoquinoxaline, 5-Bromoquinoxalin-6-amine, 6-Amino-5-bromoquinoxaline
IUPAC Name 5-bromoquinoxalin-6-amine
SMILES Nc1ccc2nccnc2c1Br
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H6BrN3/c9-7-5(10)1-2-6-8(7)12-4-3-11-6/h1-4H,10H2
Formula Weight 222.9745
Chemical Name or Material 6-Amino-5-bromoquinoxaline