missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-methylquinoline, 96%

Product Code. 15416117
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416117 250 mg 250mg
15406117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416117 Supplier Thermo Scientific Alfa Aesar Supplier No. H33298.MD
 Restricted Item
Expédition estimée: 12-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 41037-28-9 | C10H8BrN | 222.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41037-28-9
Molecular Formula C10H8BrN
Molecular Weight (g/mol) 222.085
MDL Number MFCD09258893
InChI Key XBZQSJLYRMZRSZ-UHFFFAOYSA-N
Synonym quinoline, 6-bromo-4-methyl, 6-bromo-4-methyl-quinoline, 6-bromo lepidine, 4-methyl-6-bromoquinoline, quinoline,6-bromo-4-methyl, 6-bromanyl-4-methyl-quinoline
PubChem CID 12475362
IUPAC Name 6-bromo-4-methylquinoline
SMILES CC1=C2C=C(C=CC2=NC=C1)Br

Specifications

Melting Point 94°C to 95°C
Quantity 250 mg
Sensitivity Air and light sensitive
Formula Weight 222.08
Percent Purity 96%
Chemical Name or Material 6-Bromo-4-methylquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.