Learn More
6-Chloropurine, Hydrochloride, TRC
Product Code. 30351850
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351850 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351850 Supplier Toronto Research Chemicals Supplier No. TRCC37985525G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Chloropurine, Hydrochloride
CAS 88166-54-5
Molecular Formula C5 H3 Cl N4 . Cl H
Quantity 25 g
InChI Formula InChI=1S/C5H3ClN4.ClH/c6-4-3-5(9-1-7-3)10-2-8-4;/h1-2H,(H,7,8,9,10);1H
SMILES Cl.Clc1ncnc2[nH]cnc12
IUPAC Name 6-chloro-9H-purine;hydrochloride
Molecular Weight (g/mol) 191.018
Formula Weight 189.981