Learn More
Labconco 6 ft. Nexus Horizontal Clean Bench with 21 in. Depth, Stainless Steel Interior, 230 V SC
Shop All Labconco Corporation ProductsDescription
The Nexus provides an ISO Class 4 clean airflow environment for protection of critical, non-hazardous samples.
Laminar airflow is driven by an ECM blower powered with Constant Airflow Profile (CAP™) technology for continuous, precise airflow that is energy efficient with long-lasting HEPA filters.
The sleek Nexus Horizontal Clean Bench design includes tempered safety glass and a stainless-steel interior option for effortless cleaning and durability in critical environments.
The touchscreen interface features Nexus OS™ software that simplifies operation, provides access to user settings and displays detailed alert messages when needed. Operational logs and maintenance records are stored onboard and can be accessed remotely for worry-free compliance management.
- Fully scannable 99.99+% supply HEPA filter, 99.999% efficient ULPA filter available
- Electronically Commutated Motor (ECM)
- Unidirectional, laminar airflow
- Constant Airflow Profile™ (CAP) airflow monitoring system
- Nexus Operating System and full color, 5 in. touchscreen featuring:
- Simplified home screen for easy start up and shut down operations
- Night-Smart™ for idling the blower when the system isn't in use
- Full operational, maintenance and certification logs for detailed record keeping
- Numeric and visual HEPA Filter Life graphic for easy HEPA filter monitoring
- Password security lock (optional activation) that requires user or admin password to operate the cabinet
- Status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 10% before alarming at 0%
- Interval/elapsed timer for experiment monitoring
- UV light timer (on models with UV)
- 8 languages
- Glass side panels
- Spill-Stopper design protects HEPA filter from spills
- USB port for data record export
- Remote connectivity via Ethernet port
- Prefilters to help extend HEPA filter life
- Bright, glare-free LED lighting
- Tempered safety glass and stainless steel side wall with one utility pass-through port and two service fixture punch-outs
- Electrical Outlet(s)-Ceiling mount, Flat Stainless Steel Cover
- R side on 3 and 4 ft. Models, R and L side on 5 to 8 ft. Models
- 115 V GFCI, double receptacle, maximum amperage draw 5 A
- 230 V, single receptacle, maximum amperage draw 3 A
- 30.2 in. interior height
- Optic White epoxy-coated steel
- Nominal face velocity 85 fpm (0.43 m/s)
- 10 ft. (3 m) power cord with plug
- Five year warranty
Specifications
Specifications
| Product Type | Horizontal Clean Bench |
| Coating | Epoxy |
| Filter Type | HEPA |
| Lighting Type | LED |
| Material | Stainless Steel (304) |
| Product Line | Nexus |
| Depth (English) | 21 in. |
| Depth (Metric) | 533.4 mm |
| Height (English) Interior | 30.2 in. |
| Height (Metric) Interior | 767.08 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.