Learn More
6-Methoxy-2-naphthoic Acid, TRC
Product Code. 30477105
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477105 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477105 Supplier Toronto Research Chemicals Supplier No. TRCM26475510G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Methoxynaphthalene-2-carboxylic Acid (6-Methoxy-2-naphthoic Acid; 2-Carboxy-6-methoxynaphthalene)
CAS 2471-70-7
Purity Grade Notes HPLC
Molecular Formula C12 H10 O3
Quantity 10 g
Synonym 2-Naphthalenecarboxylic acid','6-methoxy-, 2-Naphthoic acid','6-methoxy- (6CI,7CI,8CI), 6-Methoxy-2-naphthalenecarboxylic acid, 2-Methoxy-6-naphthalenecarboxylic acid, 6-Methoxy-2-naphthoic acid, 6-Methoxynaphthalen-2-carboxylic acid, NSC 408786, 2-Carboxy-6-methoxynaphthalene
InChI Formula InChI=1S/C12H10O3/c1-15-11-5-4-8-6-10(12(13)14)3-2-9(8)7-11/h2-7H,1H3,(H,13,14)
SMILES COc1ccc2cc(ccc2c1)C(=O)O
IUPAC Name 6-methoxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol) 202.21
Formula Weight 202.063
Percent Purity >95