Learn More
(+/-)-6-Methylnicotine, TRC
Product Code. 30471596
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471596 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471596 Supplier Toronto Research Chemicals Supplier No. TRCM3232555MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (+/-)-6-Methylnicotine
CAS 101540-79-8
Molecular Formula C11 H16 N2
Quantity 5 mg
Synonym Pyridine','2-methyl-5-(1-methyl-2-pyrrolidinyl)-, Pyridine','2-methyl-5-(1-methyl-2-pyrrolidinyl)-','(±)-, 2-Methyl-5-(1-methyl-2-pyrrolidinyl)pyridine, rac-6-Methylnicotine, (±)-6-Methylnicotine
InChI Formula InChI=1S/C11H16N2/c1-9-5-6-10(8-12-9)11-4-3-7-13(11)2/h5-6,8,11H,3-4,7H2,1-2H3/t11-/m0/s1
SMILES CN1CCC[C@H]1c2ccc(C)nc2
IUPAC Name 2-methyl-5-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molecular Weight (g/mol) 176.26
Formula Weight 176.1313