Learn More
7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine, 96%, BLD Pharm™
Product Code. 31031997
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
10 g
25 g
100 g
Unit Size:
10g
1g
250mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31031997 1 g 1g
31021852 100 g -
31027315 250 mg 250mg
31019867 5 g -
31023920 100 mg -
31018687 25 g -
31015785 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31031997 Supplier BLD Pharm Supplier No. BD1103491g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 105679-22-9
Molecular Formula C8H8BrNO
MDL Number MFCD09056750
Quantity 1 g
SMILES BrC1=CC2=C(C=C1)NCCO2
Molecular Weight (g/mol) 214.06
Percent Purity 96%
Chemical Name or Material 7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine