Learn More
7-Chloro-3-methyl-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine 1,1-dioxide, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 22503-72-6 |
| Molecular Formula | C8H9ClN2O2S |
| MDL Number | MFCD00270874 |
| Quantity | 250 mg |
| SMILES | O=S(C1=C(NC2C)C=CC(Cl)=C1)(N2)=O |
| Molecular Weight (g/mol) | 232.69 |
| Percent Purity | 97% |
| Chemical Name or Material | 7-Chloro-3-methyl-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine 1,1-dioxide |