missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroquinoline-4-boronic acid pinacol ester, 96%

Product Code. 15499115
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499115 5 g 5g
15489115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499115 Supplier Thermo Scientific Alfa Aesar Supplier No. H51037.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 871125-83-6 | C15H17BClNO2 | 289.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871125-83-6
Molecular Formula C15H17BClNO2
Molecular Weight (g/mol) 289.57
MDL Number MFCD07783020
InChI Key GZDUYRAAWRRVEY-UHFFFAOYSA-N
Synonym 7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloroquinoline-4-boronic acid pinacol ester, 7-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline,7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 7-chloroquinoline-4-boronic acid, pinacol ester, 7-chloroquinolin-4-yl boronic acid pinacol ester
PubChem CID 16218076
IUPAC Name 7-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
SMILES CC1(C)OB(OC1(C)C)C1=CC=NC2=CC(Cl)=CC=C12

Specifications

Quantity 5 g
Formula Weight 289.56
Percent Purity 96%
Chemical Name or Material 7-Chloroquinoline-4-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.