Learn More
JRI Maxant™ 7 Days Thermographer
Product Code. 12698241
Encompass_Preferred
Change view
Click to view available options
Time Range:
7 Days
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Time Range Unit Size
12698241 7 Days Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12698241 Supplier JRI Maxant™ Supplier No. 00059
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Specifications
Product Type 7 Days Thermographer
Diameter (Metric) 125 mm
Length (Metric) 144 mm
Quantity 1 Ea.
Temperature Range (Metric) -35°C to +15°C
Time Range 7 Days