Learn More
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline |
| CAS | 1810-74-8 |
| Molecular Formula | C13 H17 N O |
| Quantity | 1 g |
| Synonym | 1,2-Dihydro-7-methoxy-2,2,4-trimethylquinoline, 2,2,4-Trimethyl-7-methoxy-1,2-dihydroquinoline |
| InChI Formula | InChI=1S/C13H17NO/c1-9-8-13(2,3)14-12-7-10(15-4)5-6-11(9)12/h5-8,14H,1-4H3 |
| SMILES | COc1ccc2C(=CC(C)(C)Nc2c1)C |
| IUPAC Name | 7-methoxy-2,2,4-trimethyl-1H-quinoline |
| Molecular Weight (g/mol) | 203.28 |
| Formula Weight | 203.131 |