Learn More
Brady™ 7950 Series Thermal Transfer Printer Ribbons
Product Code. 10253824 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Blue
Dimensions (L x W):
300 m x 60 mm
70 m x 110 mm
70 m x 35 mm
70 m x 65 mm
70 m x 85 mm
Diameter (Metric):
12.7 mm I.D.
25.4 mm I.D.
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) Diameter (Metric) Unit Size
10253824 Black 70 m x 110 mm 12.7 mm I.D. Each
11568714 Black 300 m x 60 mm 25.4 mm I.D. Each
11508824 Black 70 m x 35 mm 12.7 mm I.D. Each
11518824 Black 70 m x 65 mm 12.7 mm I.D. Each
11874115 Blue 70 m x 110 mm 12.7 mm I.D. Each
11528824 Black 70 m x 85 mm 12.7 mm I.D. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10253824 Supplier Brady™ Supplier No. 804465
  • ONLINE EXCLUSIVE PRICE
    54.20€
    48.12€ / 1 pièces
    Économisez 6.08€
    11% de réduction
En stock
Add to basket
Edge
Add to basket
Provides strong environmental resistance against smearing and chemicals.
7950 Series Thermal Transfer Printer Ribbons
  • Sharp-edge print quality
  • Ribbon ink side out
  • Wax/resin ribbon formulation
Specifications
Product Type Thermal Transfer Printer Ribbon
Color Black
Diameter (Metric) 12.7 mm I.D.
Dimensions (L x W) 70 m x 110 mm
Description Wax/Resin ribbon formulation
Printable Sides Ink side out
Alternate Part Number R7950-110X70/O