Learn More
(7R,11R,E)-3,7,11,15-Tetramethylhexadec-2-en-1-ol, 95%, BLD Pharm™
Product Code. 30782790
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30782790 1 g 1g
30774811 10 g 10g
30770834 25 g 25g
30776855 500 g -
30780607 5 g 5g
30778710 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30782790 Supplier BLD Pharm Supplier No. BD1479771g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 150-86-7
Molecular Formula C20H40O
MDL Number MFCD00151280
Quantity 1 g
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC/C(C)=C/CO
Molecular Weight (g/mol) 296.53
Percent Purity 95%
Chemical Name or Material (7R,11R,E)-3,7,11,15-Tetramethylhexadec-2-en-1-ol