Learn More
8-Hydroxy-2-methyl-4(3H)-quinazolinone, TRC
Product Code. 30397536
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397536 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397536 Supplier Toronto Research Chemicals Supplier No. TRCH94833010MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 8-Hydroxy-2-methyl-4(3H)-quinazolinone
CAS 90417-38-2
Molecular Formula C9 H8 N2 O2
Quantity 10 mg
Synonym 4(3H)-Quinazolinone, 8-hydroxy-2-methyl-, 4(1H)-Quinazolinone, 8-hydroxy-2-methyl- (9CI), 8-Hydroxy-2-methyl-4(3H)-quinazolinone, NSC 696807, Nu 1025
InChI Formula InChI=1S/C9H8N2O2/c1-5-10-8-6(9(13)11-5)3-2-4-7(8)12/h2-4,12H,1H3,(H,10,11,13)
SMILES CC1=Nc2c(O)cccc2C(=O)N1
IUPAC Name 8-hydroxy-2-methyl-3H-quinazolin-4-one
Molecular Weight (g/mol) 176.17
Formula Weight 176.0586