Learn More
9-cis-Retinoic Acid, TRC
Product Code. 18031720
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18031720 25 mg 25mg
30134331 5 mg 5mg
18081220 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18031720 Supplier Toronto Research Chemicals Supplier No. TRCR24500025MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5300-03-8
Molecular Formula C20 H28 O2
Molecular Weight (g/mol) 300.44
Synonym 9-cis-Retinoic Acid, Retinoic acid, cis-9,trans-13- (8CI), 9-cis-Retinoic acid, 9(Z)-Retinoic acid, 9-cis-Tretinoin, AGN 192013, ALRT 1057, Alitretinoin, LG 100057, LGD 100057, LGD 1057, NSC 659772, Panretin, Panretyn, Panrexin, Toctino, BAL0004079-000, code for free acid, BAL4079, 9-cis retinoic acid, 9-(Z) Retinoic Acid, R0004079-000, CRA-Alitretinoin, INN Alitretinoin
IUPAC Name (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES C\C(=C\C=C\C(=C\C(=O)O)\C)\C=C\C1=C(C)CCCC1(C)C
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C20H28O2/c1-15(8-6-9-16(2)14-19(21)22)11-12-18-17(3)10-7-13-20(18,4)5/h6,8-9,11-12,14H,7,10,13H2,1-5H3,(H,21,22)/b9-6+,12-11+,15-8-,16-14+
Formula Weight 300.2089
Chemical Name or Material 9-cis-Retinoic acid