Learn More
uvex 9906 Pheos Replacement Visor Lens
Product Code. 17220282 Shop All Uvex Products
Change view
Click to view available options
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17220282 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17220282 Supplier uvex Supplier No. 9906013
  • ONLINE EXCLUSIVE PRICE
    33.65€
    31.21€ / 1 pièces
    Économisez 2.44€
    7% de réduction
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Replacement visor lens for pheos series safety helmets is compatible with model 9906.002 (mechanical lever arms), or model 9906.003 (self-locking magnetic catches).
  • Replacement lens for uvex pheos model 9906.002 (mechanical lever arms) or model 9906.003 (self-locking magnetic catches)
  • Lightweight, robust face shield for uvex pheos industrial safety helmet ES-WR / BS-WR and uvex pheos alpine (visor adapter 9790.067 required)
  • Labeling: 2C-1.2 W1 B 9 KN
  • Features supravision excellence coating
Supravision Excellence Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant and chemical-resistant
  • Easy to clean, and less susceptible to soiling
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 170:2002
For Use With (Equipment) Pheos 9906.002, 9906.003
Lens Tint Clear
Material Polycarbonate