Learn More
Abcam Anti-Human IgG4 pFc' [HP6023] (FITC), 250 ug
Product Code. 32116179 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116179 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116179 Supplier Abcam Supplier No. ab99815250ug
 Restricted Item
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Mouse Monoclonal antibody, suitable for Dot, ELISA, WB, ICC/IF, with 2 citations.
  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG3
  • Applications: Dot, ELISA, WB, ICC/IF
  • Species Reactivity: Human
  • Storage Buffer: pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein A
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Applications Dot Blot, ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone HP6023
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
Gene IGHG4
Gene Symbols IGHG4
Host Species Mouse
Purification Method Affinity purification Protein A
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG3
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Target Molecule IGHG4, Human
Validated Application abrv Dot, ELISA, WB, ICC/IF