Learn More
Abcam Rabbit Anti-Goat IgG H&L (Glucose Oxidase), 500 uL
Product Code. 32108337 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32108337 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32108337 Supplier Abcam Supplier No. ab136719500uL
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Rabbit Anti-Goat IgG H&L (Glucose Oxidase)
  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ELISA, ICC, IHC-Fr, IHC-P, WB
  • Species Reactivity: Goat
  • Storage Buffer: Constituents: PBS, 50% Glycerol (glycerin, glycerine)
  • Form: Liquid
  • Purification: Affinity purification Immunogen
Specifications
Applications Western Blot, ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Formulation Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Host Species Rabbit
Purification Method Affinity purification Immunogen
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Goat.
Target Species Goat
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Gt
Validated Application abrv WB, ELISA, ICC, IHC-P, IHC-Fr