Learn More
Abcam Rat monoclonal Anti-Mouse IgG2a heavy chain (HRP) [LO-MG2a-9], 100 ug
Product Code. 32117075 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32117075 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32117075 Supplier Abcam Supplier No. ab11571100ug
 Restricted Item
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Rat Monoclonal antibody, suitable for ELISA, with 3 citations.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Applications ELISA
Classification Monoclonal
Clone LO-MG2a-9
Conjugate Unconjugated
Formulation Preservative: 0.01% Thimerosal (merthiolate), Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene Mouse IgG2a
Host Species Rat
Purification Method Affinity purification Protein G
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Mouse IgG2a
Validated Application abrv ELISA