Learn More
Thermo Scientific™ Abgene™ SmartCap Mats
GREENER_CHOICE
Product Code. 12463847
Change view
Click to view available options
No. of Wells:
96
Unit Size:
Pack of 960
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Unit Size
12463847 96 Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12463847 Supplier Thermo Scientific™ Supplier No. AB1413
  • ONLINE EXCLUSIVE PRICE
    178.00€
    155.00€ / 960 pièces
    Économisez 23.00€
    12% de réduction
Expédition estimée: 22-02-2027
Add to basket
Add to basket
Bulk Quote Request
This item is not returnable. View return policy
The Thermo Scientific™ Abgene™ SmartCap Mat ensures reliable sealing from ambient temperatures down to -80 °C.
The Thermo Scientific™ Abgene™ SmartCap Mat seals 96 samples at one time in a high-density storage format,affording maximum space saving benefits. Suitable for use with Thermo Scientific storage tubes ranging from 0.3 ml to1.4 ml, the Abgene SmartCap Mat maintain sa consistent seal down to -80°C.
Features:
  • Efficient Storage
  • Enhanced Materials
  • Ease of Installation and Access
TRUSTED_SUSTAINABILITY
Specifications
Description Seals 96 sample tubes at one time
No. of Wells 96
Product Type Sealing Mat
Temperature Range (Metric) -80°C
Green Features ACT Ecolabel (Silver)