Learn More
3M™ Doodlebug™ Abrasive Sponge 8242
Product Code. 17188850
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17188850 Blue Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17188850 Supplier 3M™ Doodlebug™ Supplier No. 7100194280
  • ONLINE EXCLUSIVE PRICE
    259.00€
    146.46€ / 30 pièces
    Économisez 112.54€
    43% de réduction
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For medium duty cleaning tasks such as cleaning stairs
  • Reduces rolls to eliminate scratches on the floor
  • Removes dirt and scratches from various surfaces
  • For moderately aggressive cleaning
  • Easy to clean after use
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Dimensions (L x W) 118 x 254 mm
For Use With (Application) Medium Duty Cleaning Tasks