Learn More
Ac-L-Gln-OH, 97%, BLD Pharm™
Product Code. 30945259
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
10 g
25 g
100 g
500 g
Unit Size:
100g
100mg
10g
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30945259 25 g 25g
30936943 100 g 100g
30941558 100 mg 100mg
30934854 10 g 10g
30944166 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30945259 Supplier BLD Pharm Supplier No. BD14302725g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 2490-97-3
Molecular Formula C7H12N2O4
MDL Number MFCD00038159
Quantity 25 g
SMILES O=C(O)[C@@H](NC(C)=O)CCC(N)=O
Molecular Weight (g/mol) 188.18
Percent Purity 97%
Chemical Name or Material Ac-L-Gln-OH