Learn More
Fisherbrand™ accumet™ AR Series Meter-Attached Electrode Holder
Product Code. 31702809 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
Holder Attachment
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
31702809 Holder Attachment 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 31702809
  • ONLINE EXCLUSIVE PRICE
    228.00€
    130.47€ / 1 kits
    Économisez 97.53€
    42% de réduction
Expédition estimée: 09-12-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
For use with AR series meters
  • Accessory designed for use with AR series meters, securely holds electrodes during measurements for accurate and stable readings
TRUSTED_SUSTAINABILITY
Specifications
Type Holder Attachment
For Use With (Equipment) AR Series Meters