Learn More
Acenaphthylene, TRC
Product Code. 30317203
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317203 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317203 Supplier Toronto Research Chemicals Supplier No. TRCA130750100G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Acenaphthylene
CAS 208-96-8
Molecular Formula C12 H8
Quantity 100 g
Synonym Acenaphthylene, Cyclopenta[de]naphthalene, NSC 59821, acenaphthyene
InChI Formula InChI=1S/C12H8/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-8H
SMILES C1=Cc2cccc3cccc1c23
IUPAC Name acenaphthylene
Molecular Weight (g/mol) 152.19
Formula Weight 152.0626