Learn More
Thermo Scientific™ AcroMetrix CT/NG Control (IVD, CE-IVD)
Product Code. 17848581 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17848581 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17848581 Supplier Thermo Scientific™ Supplier No. 967146
 Restricted Item
Expédition estimée: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Assess the performance of nucleic acid test procedures for the qualitative determination of CT and NG nucleic acids with this control, which is for In Vitro Diagnostic Use.
Assess the performance of nucleic acid test procedures for the qualitative determination of Chlamydia trachomatis (CT) and Neisseria gonorrhoeae (NG) nucleic acids in swab and urine patient specimens with Thermo Scientific™ AcroMetrix™ CT/NG Control. For verification of an effective nucleic acid extraction procedure, the AcroMetrix CT/NG Control contains intact, inactivated C. trachomatis and N. gonorrhoeae. Routine use enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation. The control is for In Vitro Diagnostic Use.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Chlamydia trachomatis (CT) nucleic acids, Neisseria gonorrhoeae (NG) nucleic acids
Type CT/NG Control
Packaging 20mL
Quantity 20 mL